missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 Antibody (4E11B11) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80702
This item is not returnable.
View return policy
Description
DRP1 Monoclonal specifically detects DRP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DRP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:500 - 1:2000, ELISA : 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin : 1:200 - 1:1000 | |
| DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4E11B11 | |
| Unconjugated | |
| PBS | |
| DNM1L | |
| Partial recombinant human DRP1 (between residues 100-250) expressed in E. coli. [Uniprot: O00429] | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction | |
| 10059 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction