missing translation for 'onlineSavingsMsg'
Learn More

DRP1 Antibody (4E11B11), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18432915 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432915 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432915 Supplier Novus Biologicals Supplier No. NBP280702AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DRP1 Monoclonal antibody specifically detects DRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4E11B11
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1
Host Species Mouse
Immunogen Partial recombinant human DRP1 (between residues 100-250) expressed in E. coli. [Uniprot: O00429]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10059
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.