missing translation for 'onlineSavingsMsg'
Learn More

Drotaveraldine, TRC

Product Code. 17952095
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17952095 100 mg 100mg
17950855 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17952095 Supplier Toronto Research Chemicals Supplier No. TRCD689670100MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54088-62-9
Molecular Formula C24H29NO5
Molecular Weight (g/mol) 411.49
Synonym Methanone, (6,7-diethoxy-3,4-dihydro-1-isoquinolinyl)(3,4-diethoxyphenyl)-, (6,7-Diethoxy-3,4-dihydro-1-isoquinolinyl)(3,4-diethoxyphenyl)methanone, Dihydroperparaldine, Drotaveraldine
SMILES CCOc1ccc(cc1OCC)C(=O)C2=NCCc3cc(OCC)c(OCC)cc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H29NO5/c1-5-27-19-10-9-17(14-20(19)28-6-2)24(26)23-18-15-22(30-8-4)21(29-7-3)13-16(18)11-12-25-23/h9-10,13-15H,5-8,11-12H2,1-4H3
Formula Weight 411.2046
Percent Purity >95%
Chemical Name or Material Drotaveraldine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.