missing translation for 'onlineSavingsMsg'
Learn More

Drospirenone, MedChemExpress

Product Code. 16494237
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494237 50 mg 50mg
16468348 10 mg 10mg
16438106 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494237 Supplier Medchem Express Supplier No. HYB0111
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Drospirenone(Dihydrospirorenone) is a synthetic progestin that is an analog to spironolactone.

Drospirenone(Dihydrospirorenone) is a synthetic progestin that is an analog to spironolactone.

Specifications

Chemical Name or Material Drospirenone
CAS 67392-87-4
Color White
Purity Grade Notes Research
Molecular Formula C24H30O3
Quantity 50 mg
Synonym Dihydrospirorenone
Solubility Information DMSO : 50 mg/mL (136.43 mM; Need ultrasonic)
SMILES C[C@@]12[C@](OC3=O)(CC3)[C@@H](C4)[C@@H]4[C@@]1([H])[C@@]([C@@H]5[C@H]6C5)([H])[C@]([C@](C6=CC7=O)(CC7)C)([H])CC2
Molecular Weight (g/mol) 366.49
Formula Weight 366.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.