Learn More
Invitrogen™ Drosha Recombinant Rabbit Monoclonal Antibody (JU33-01)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532818
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Members of the ribonuclease III superfamily of double-stranded (ds) RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells (Fortin et al., 2002). The RNase III Drosha is the core nuclease that executes the initiation step of microRNA (miRNA) processing in the nucleus (Lee et al., 2003).
Specifications
| Drosha | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NRR4 | |
| DROSHA | |
| Synthetic peptide within Human Drosha aa 30-79. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| JU33-01 | |
| Unconjugated | |
| DROSHA | |
| 1110013A17Rik; AI874853; Drosha; drosha ribonuclease III; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; ethanol induced 2; ETOHI2; HSA242976; MGC115770; nuclear RNase III Drosha; OTTHUMP00000220910; OTTHUMP00000220911; OTTHUMP00000220912; OTTHUMP00000220913; p241; Protein Drosha; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; RANSE3L; RGD1307626; ribonuclease 3; Ribonuclease III; ribonuclease III, nuclear; ribonuclease type III, nuclear; RN3; RNase III; RNASE3L; RNASEN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29102 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.