missing translation for 'onlineSavingsMsg'
Learn More

Droperidol

Product Code. 15638695 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15638695

Brand: Thermo Scientific Acros 460840010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 548-73-2 | C22H22FN3O2 | 379.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 548-73-2
Melting Point 148.0°C to 149.0°C
Loss on Drying 5% max. (70°C, 4 h) (vaccuum)
Infrared Spectrum Authentic
Molecular Formula C22H22FN3O2
Quantity 1 g
Synonym droperidol, droleptan, inapsine, dehydrobenzperidol, properidol, sintodril, sintosian, inapsin, vetkalm, dridol
Solubility Information Solubility in water: .. Other solubilities: soluble in chlorofom
InChI Key RMEDXOLNCUSCGS-UHFFFAOYSA-N
SMILES C1CN(CC=C1N2C3=CC=CC=C3NC2=O)CCCC(=O)C4=CC=C(C=C4)F
IUPAC Name 3-[1-[4-(4-fluorophenyl)-4-oxobutyl]-3,6-dihydro-2H-pyridin-4-yl]-1H-benzimidazol-2-one
Molecular Weight (g/mol) 379.43
PubChem CID 3168
ChEBI CHEBI:4717
Formula Weight 379.43
Chemical Name or Material Droperidol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.