missing translation for 'onlineSavingsMsg'
Learn More

Drofenine Hydrochloride, TRC

Product Code. 30380881
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380881 500 mg
30371828 100 mg
30383295 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380881 Supplier Toronto Research Chemicals Supplier No. TRCD679000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-66-3
Molecular Formula C20 H31 N O2 . Cl H
Molecular Weight (g/mol) 353.93
Synonym Drofenine Hydrochloride, α-Cyclohexyl-benzeneacetic Acid 2-(Diethylamino)ethyl Ester Hydrochloride, α-Phenyl-cyclohexaneacetic Acid 2-(Diethylamino)ethyl Ester Hydrochloride, 2-(Diethylamino)ethyl α-Cyclohexylbenzeneacetate Hydrochloride, Adiphenine H hydrochloride, Cycloadiphenine, Cycloadiphenine hydrochloride, Cyclospasmol [Tempelhof], Cyclovegantine, Drofenine Hydrochloride, Hexahydroadiphenine Hydrochloride, IT 19, NSC 42559, Trasentin H, Trasentine A
IUPAC Name 2-(diethylamino)ethyl 2-cyclohexyl-2-phenylacetate;hydrochloride
SMILES Cl.CCN(CC)CCOC(=O)C(C1CCCCC1)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H31NO2.ClH/c1-3-21(4-2)15-16-23-20(22)19(17-11-7-5-8-12-17)18-13-9-6-10-14-18;/h5,7-8,11-12,18-19H,3-4,6,9-10,13-16H2,1-2H3;1H
Formula Weight 353.2122
Chemical Name or Material Drofenine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.