missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drofenine hydrochloride, MedChemExpress
Description
Drofenine hydrochloride is a potent competitive inhibitor of BChE, and the ki values of Drofenine is calculated to be 3 uM.
Specifications
Specifications
| Chemical Name or Material | Drofenine hydrochloride |
| CAS | 548-66-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H32ClNO2 |
| Quantity | 50 mg |
| Synonym | Hexahydroadiphenine hydrochloride |
| Solubility Information | DMSO : ≥ 75 mg/mL (211.91 mM) ∣H2O : ≥ 33.3 mg/mL (94.09 mM) |
| SMILES | O=C(OCCN(CC)CC)C(C1CCCCC1)C2=CC=CC=C2.[H]Cl |
| Molecular Weight (g/mol) | 353.93 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?