missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, With indicator, 4 mesh; Honeywell™ Fluka™

Product Code. 15632400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
454 g
2.3 kg
Packaging:
Glass Bottle
Unit Size:
2.3kg
454g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15632400 454 g Glass Bottle 454g
15622400 2.3 kg Glass Bottle 2.3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15632400 Supplier Honeywell-Fluka Supplier No. 238961454G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With indicator, 4 mesh, Drierite™ is a trademark of W. A. Hammond Drierite Company, Ltd.

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Material Calcium Sulfate
Quantity 454 g
Linear Formula CaSO4
Mesh Size 4mesh
UN Number UN3077
Packaging Glass Bottle
IUPAC Name calcium;sulfate
Formula Weight 136.14g/mol
Chemical Name or Material Calcium Sulfate, With indicator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.