missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, indicating (CaSO{4}), Thermo Scientific™

Product Code. 15411867
Change view
Click to view available options
Quantity:
2270 g
Unit Size:
2270g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411867 2270 g 2270g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15411867 Supplier Thermo Scientific Alfa Aesar Supplier No. 043064.=A
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Blue
Physical Form Granular
CAS 7778-18-9
Quantity 2270 g
Molecular Formula CaO4S
MDL Number MFCD00010912
Odor Odorless
Solubility Information Soluble aq. soln..
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES [Ca++].[O-]S([O-])(=O)=O
Sensitivity Hygroscopic
IUPAC Name calcium sulfate
Molecular Weight (g/mol) 136.13
PubChem CID 24497
ChEBI CHEBI:31346
Grade Laboratory
Chemical Name or Material Drierite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.