missing translation for 'onlineSavingsMsg'
Learn More

PSF Progressive Safety™ Dri-Force Plus Black PVC Safety Wellington Boots

Product Code. 15868366
Click to view available options
Size:
3
4
5
6
7
8
9
10
11
12
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15868366

Brand: PSF Progressive Safety™ D110SM12

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A rugged and stylish safety footwear solution, these safety Wellingtons provide 200 joules toe protection.

  • All safety footwear is designed with both comfort and durability in mind
  • Conforms to EN ISO 20345:2011
  • Manufactured from black S5 PVC
  • 100% waterproof
  • PVC upper, steel midsole, PVC sole
  • Antistatic
  • SRC rated

Specifications

Certifications/Compliance EN ISO 20345:2011
Color Black
Design Wellington boot
For Use With (Application) Safety
Material PVC
Size 12
Waterproof Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.