missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRI™ Endocrine Calibrator Sets

Product Code. 17734618 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17734618 6 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17734618 Supplier Thermo Scientific™ Supplier No. 0476
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve excellent workflow management, cost savings and superior performance with these endocrine calibrator sets.

Achieve excellent workflow management, cost savings and superior performance with Thermo Scientific™ DRI™ Endocrine Calibrator Sets. Packaged in convenient sizes with applications on a multitude of clinical chemistry analyzers, Endocrine Assays T4 and T Uptake are available in both Thermo Scientific™ CEDIA™ and DRI formats.

  • Packaged in convenient sizes
  • Applications on a multitude of clinical chemistry analyzers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Total Thyroxine (T4) Calibrator Kit
Detectable Analytes Negative Calibrator, Thyroxine, Thyroxine
Sample Type Plasma, Serum
Type Calibrator
Quantity 6 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.