missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRI™ Endocrine Assays

Product Code. 17730308 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL, 34 mL
Type:
Total Thyroxine Assay
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Type Unit Size
17730308 100 mL, 34 mL Total Thyroxine Assay Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17730308 Supplier Thermo Scientific™ Supplier No. 0454
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve excellent workflow management, cost savings and superior performance with DRI Endocrine Assays

Achieve excellent workflow management, cost savings and superior performance with Thermo Scientific™ DRI™ Endocrine Assays, packaged in convenient sizes with applications on a multitude of clinical chemistry analyzers. Endocrine Assays T4 and T Uptake are available in both DRI and Thermo Scientific™ CEDIA™ formats.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Thyroxine
Sample Type Plasma, Serum
Type Total Thyroxine Assay
Quantity 100 mL, 34 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.