missing translation for 'onlineSavingsMsg'
Learn More

Techne™ Dri-Block™ Aluminum Heating Block Insert

Catalog Number 15565760
Click to view available options
(12 x 16 mm)
This item is not returnable. View return policy
This item is not returnable. View return policy

Techne™ Dri-Block™ Aluminum Heating Block Inserts are precision-engineered and offer excellent temperature accuracy

  • Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates
  • Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance
  • Most blocks come with a separate hole to accept a thermometer
  • Precision-engineered aluminum heating blocks offer excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
Capacity (English) 12 Block Openings
Material Aluminum
Type Flat-Bottom
Tube Size 16 mm
Dimensions (L x W) (12 x 16 mm)
Depth (Metric) 25 mm/Opening
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'