missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit
Shop All Applied Biosystems ProductsDescription
Includes
Each kit contains dRhodamine terminator ready reaction mix, M13 (-21) primer, pGEM control DNA, and a user's manual.
- Increase your signal-to-noise ratio
- Simplify workflows with easy-to-clean-up chemistries
- Get optimized chemistry for sequencing templates with homo-polymers (AA's, TT's, etc.)
- Obtain clean signal, even peak patterns, and minimal mobility shifts with improved dye terminators
- Reduce spectrum overlap with narrow dye emission spectra
Specifications
Specifications
| Content And Storage | Each kit contains dRhodamine Terminator Ready Reaction Mix, M13 (-21) Primer, pGEM Control DNA, and a user's manual. There are sufficient reagents for 1000 reactions. Store kit at -15 to -25°C and avoid repeated freeze-thaw cycles. |
| Format | 8-well Strip, 96-well Plate |
| For Use With (Equipment) | 310 Genetic Analyzer, 3130xl Genetic Analyzer, ABI PRISM™ 377, GeneAmp 2700, GeneAmp 9600, GeneAmp 9700, GeneAmp™ 2400, 3130 Genetic Analyzer |
| For Use With (Application) | Standard Sequencing |
| Includes | dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit, 1000 rxns, 8-well Strips, 96-well Plate, For 310 Genetic Analyzer, 3130 Genetic Analyzer |
| No. of Reactions | 1000 Reactions |
| Product Type | Sequencing Kit |
| Quantity | 1,000 reactions |
| Technique | Fluorescent Dye Terminator Sequencing |
| Template Compatibility | Single Stranded DNA, Plasmid DNA (≤15Kb), PCR Amplicons |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?