missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit

Product Code. 15869736 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15869736 100 Reactions Each
15879736 1000 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15869736 Supplier Applied Biosystems™ Supplier No. 403044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Each kit contains 800uL tube of dRhodamine terminator ready reaction mix, 1 tube M13 (-21) primer, 1 tube pGEM control DNA, protocol manual.

The dRhodamine Terminator Cycle Sequencing Kit provides optimized dRhodamine dideoxy terminator chemistries for decreased background noise in a wide array sequencing applications.

  • Increase your signal-to-noise ratio
  • Simplify workflows with easy-to-clean-up chemistries
  • Get optimized chemistry for sequencing templates with homo-polymers (AA's, TT's, etc.)
  • Obtain clean signal, even peak patterns, and minimal mobility shifts with improved dye terminators
  • Reduce spectrum overlap with narrow dye emission spectra
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each kit contains 800uL Tube of dRhodamine Terminator Ready Reaction Mix, 1 Tube M13 (-21) Primer, 1 Tube pGEM Control DNA, Protocol Manual. Store kit at -15 to -25°C and avoid repeated freeze-thaw cycles.
Format 8-well Strip, 96-well Plate
For Use With (Equipment) 310 Genetic Analyzer, 3130xl Genetic Analyzer, ABI PRISM™ 377, GeneAmp 2700, GeneAmp 9600, GeneAmp 9700, GeneAmp™ 2400, 3130 Genetic Analyzer
For Use With (Application) Standard Sequencing
Includes dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit, 100 rxns, 8-well Strips, 96-well Plate, For 310 Genetic Analyzer, 3130 Genetic Analyzer
No. of Reactions 100 Reactions
Product Type Sequencing Kit
Quantity 100 reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility Single Stranded DNA, Plasmid DNA (≤15Kb), PCR Amplicons

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.