missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549155
This item is not returnable.
View return policy
Description
Predicted to react with bovine based on sequence homology.
3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
Specifications
| DRG2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| P55039, Q9QXB9 | |
| Drg2 | |
| KLH conjugated synthetic peptide between 154-180 amino acids from the Central region of human DRG2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Drg2 | |
| AI255295; AI461605; developmentally regulated GTP binding protein 2; developmentally regulated GTP-binding protein 2; developmentally-regulated GTP-binding protein 2; DRG2; DRG-2; RGD1562380; TRAFAC GTPase DRG2; Translation factor GTPase DRG2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 13495, 1819 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction