missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DRG2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17137593
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137593 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17137593 Supplier Invitrogen™ Supplier No. PA5109661

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TKGEVQRSLL EKELESVGIR LNKHKPNIYF KPKKGGGISF NSTVTLTQCS EKLVQLILHE YKIFNAEVLF REDCSPDEFI DVIVGNRVYM PCLYVYNKID QISMEEVDRL A.

3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRG2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Drg2
Gene Accession No. P55039, Q9QXB9
Gene Alias AI255295; AI461605; developmentally regulated GTP binding protein 2; developmentally regulated GTP-binding protein 2; developmentally-regulated GTP-binding protein 2; DRG2; DRG-2; RGD1562380; TRAFAC GTPase DRG2; Translation factor GTPase DRG2
Gene Symbols Drg2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13495, 1819, 497915
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.