missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£222.00 - £470.00
Specifications
| Antigen | DRG2 |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18464201
|
Novus Biologicals
NBP1-89461-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18249026
|
Novus Biologicals
NBP1-89461 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DRG2 Polyclonal specifically detects DRG2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DRG2 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1819 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:YKIFNAEVLFREDCSPDEFIDVIVGNRVYMPCLYVYNKIDQISMEEVDRLARKPNSVVISCGMKLNLDYLLEMLWEYLALTCIYTKKRGQRPDFTDAIILRKGASVEHVCHRIHRSLASQFKYALVWGTSTKYSPQRVGLTHTMEHEDVI | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| developmentally regulated GTP binding protein 2, developmentally regulated GTP-binding protein 2, developmentally-regulated GTP-binding protein 2, DRG-2 | |
| DRG2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title