missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRG1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13190-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with domit-negative AES. Unusual function as coactivator for ESRRG.Specifications
| DRG1 | |
| Polyclonal | |
| Unconjugated | |
| DRG1 | |
| DRG 1, DRG1, NEDD 3, NEDD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305470, 4733 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y295 | |
| DRG1 | |
| DRG1 Fusion Protein Ag3866 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction