missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DRG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550398
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total DRG1 protein.
Catalyzes the conversion of GTP to GDP through hydrolysis of the gamma-phosphate bond in GTP (PubMed:29915238, PubMed:23711155). Appears to have an intrinsic GTPase activity that is stimulated by ZC3H15/DFRP1 binding likely by increasing the affinity for the potassium ions (PubMed:23711155). When hydroxylated at C-3 of 'Lys-22' by JMJD7, may bind to RNA and play a role in translation (PubMed:19819225, PubMed:29915238). Binds to microtubules and promotes microtubule polymerization and stability that are required for mitotic spindle assembly during prophase to anaphase transition. GTPase activity is not necessary for these microtubule-related functions (PubMed:28855639). [UniProt].
Specifications
| DRG1 | |
| Polyclonal | |
| Unconjugated | |
| DRG1 | |
| AA408859; AI132520; developmentally regulated GTP binding protein 1; developmentally regulated GTP-binding protein 1; developmentally-regulated GTP-binding protein 1; Drg; Drg1; DRG-1; GTP-binding protein; NEDD3; NEDD-3; neural precursor cell expressed developmentally down-regulated protein 3; neural precursor cell expressed, developmentally down-regulated 3; TRAFAC GTPase DRG1; Translation factor GTPase DRG1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13494, 4733 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P32233, Q9Y295 | |
| DRG1 | |
| Fusion protein corresponding to residues near the C terminal of human developmentally regulated GTP binding protein 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction