missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRG1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92437-0.1ml
This item is not returnable.
View return policy
Description
DRG1 Polyclonal antibody specifically detects DRG1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DRG1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| developmentally regulated GTP binding protein 1, developmentally regulated GTP-binding protein 1, developmentally-regulated GTP-binding protein 1, DKFZp434N1827, DRG-1, NEDD3, NEDD-3, Neural precursor cell expressed developmentally down-regulated protein 3, neural precursor cell expressed, developmentally down-regulated 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 158-367 of human DRG1 (NP_004138.1). KKIIENELEGFGIRLNSKPPNIGFKKKDKGGINLTATCPQSELDAETVKSILAEYKIHNADVTLRSDATADDLIDVVEGNRVYIPCIYVLNKIDQISIEELDIIYKVPHCVPISAHHRWNFDDLLEKIWDYLKLVRIYTKPKGQLPDYTSPVVLPYSRTTVEDFCMKIHKNLIKEFKYALVWGLSVKHNPQKVGKDHTLEDEDVIQIVKK | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
| 4733 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction