missing translation for 'onlineSavingsMsg'
Learn More

Dreyers Agglutination Tubes

Product Code. 13048423
Click to view available options
Diameter (Metric):
7 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 13048423

Brand: S Murray SAMCO G115/21

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Length (Metric) 60 mm
Material Borosilicate Glass
Packaging 100
Quantity 100 Pack
Diameter (Metric) 7 mm
Product Type Dreyers Agglutination Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.