missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DREF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76327
This item is not returnable.
View return policy
Description
DREF Polyclonal specifically detects DREF in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| DREF | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9189 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000 | |
| AAH15030 | |
| ZBED1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human ZBED1. The immunogen is located within amino acids 180 - 230 of ZBED1. Amino Acid Squence: STDMWRSENQNRAYV | |
| 0.1 mg | |
| Primary | |
| This antibody is predicted to have no cross-reactivity to ZBED2 or ZBED3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction