Learn More
Lenz Laborglasintrumente™ Drechsel Gas Washing Bottle without Filter Disc
Product Code. 10391760
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10391760 250 mL Each
1 options
Product Code. 10391760 Supplier Lenz Laborglasintrumente™ Supplier No. 05500512
Only null left
Add to basket
Add to basket
Gas washing bottles acc. to Drechsel, without filter disc, complete with head
  • Acc. to DIN 12596
  • Complete with head
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
Capacity (Metric) 250 mL
Type Thread GL 14
Quantity 1