missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Drebrin Recombinant Rabbit Monoclonal Antibody (23GB5145), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Drebrin |
|---|---|
| Clone | 23GB5145 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089455
![]() |
Invitrogen™
MA552804 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088774
|
Invitrogen™
MA552805 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Drebrin Recombinant Monoclonal antibody specifically detects Drebrin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Drebrin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q07266, Q16643, Q9QXS6 | |
| 1627, 56320, 81653 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5145 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D0S117E; Dbn1; developmentally regulated brain protein; developmentally-regulated brain protein; DKFZp434D064; Drba; Drebrin; drebrin 1; drebrin A; drebrin E; drebrin E2; drebrin-1; Unknown (protein for MGC:138067) | |
| A synthesized peptide derived from human Drebrin (120-181AA). | |
| Primary | |
| -20°C | |
| DBN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title