missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drebrin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25770-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer′s disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.Specifications
| Drebrin | |
| Polyclonal | |
| Unconjugated | |
| DBN1 | |
| D0S117E, DBN1, Drebrin, drebrin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1627, 56320 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16643, Q9QXS6 | |
| Dbn1 | |
| Drebrin Fusion Protein Ag22634 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction