missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
DreamTaq Hot Start DNA Polymerase; 10X DreamTaq Green Buffer
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.
Better reaction outcomes:
Fidelity (vs. Taq): 1X
Better reaction outcomes:
- Higher yield of target amplicons from low template amounts.
- Increased reaction specificity due to reliable hot-start technology.
- Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
- Room temperature reaction set-up.
- Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.
- Direct loading of PCR products on a gel with DreamTaq Green buffer.
Fidelity (vs. Taq): 1X
Recommended Storage
Store at -20C.
Specifications
Specifications
| Content And Storage | Contains: • 40 μL DreamTaq Hot Start DNA Polymerase (5 U/μL) • 1.25 mL 10X DreamTaq Green Buffer (includes 20 mM MgCl2) Store at -5 to -30°C. |
| GC-Rich PCR Performance | Low |
| Polymerase | DreamTaq DNA Polymerase |
| Reaction Speed | Standard |
| Quantity | 200 U |
| Shipping Condition | Wet or Dry Ice |
| Hot Start | Built-In Hot Start |
| Overhang | 3'-A |
| Reaction Format | Separate Components |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?