missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DREADD agonist 21

Product Code. 18065292
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18065292

Brand: Tocris Bioscience™ 5548/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent hM3Dq and hM4Di DREADD agonist; blood brain barrier penetrant

DREADD agonist 21 is a potent muscarinic DREADD agonist (pEC50 values are 8.91, 8.48 and 7.77 for hM1Dq, hM3Dq and hM4Di in vitro, respectively). Exhibits > 10-fold higher affinity for hM1Dq and hM4Di compared to wild type receptors. Also displays little to no activity at wild type M3 receptors. Activates neurons expressing hM3Dq DREADDS and inhibits activity in neurons expressing hM4Di in vivo. Displays excellent brain permeability. Water soluble salt also available, DREADD agonist 21 hydrochloride (Cat. No. 6422).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-(1-Piperazinyl)-5H-dibenzo[b,e][1,4]diazepine
CAS 56296-18-5
Quantity 50 mg
Target DREADD Activators
Molecular Formula C17H18N4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.