missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DRD5 Recombinant Rabbit Monoclonal Antibody (HL3081)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556686
This item is not returnable.
View return policy
Description
DRD5 Recombinant Monoclonal Antibody for ICC/IF, IHC (P)
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
Specifications
| DRD5 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P21918, Q8BLD9 | |
| DRD5 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human Dopamine Receptor D5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| HL3081 | |
| Unconjugated | |
| DRD5 | |
| D(1B) dopamine receptor; D(5) dopamine receptor; D1B; D1b receptor; D1beta dopamine receptor; D5R; DBDR; Dopamine 5 receptor; dopamine D5 receptor; dopamine receptor 5; dopamine receptor D1B; dopamine receptor D5; DRD 1B; DRD 5; Drd1b; DRD1L2; Drd5; Drd-5; Gpcr1; Hd5dr; MGC10601 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13492, 1816 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction