missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ D4 Dopamine Receptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572569
This item is not returnable.
View return policy
Description
D4 Dopamine Receptor Polyclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes the D4 subtype of the dopamine receptor. The D4 subtype is a G-protein coupled receptor which inhibits adenylyl cyclase. It is a target for drugs which treat schizophrenia and Parkinson disease. Mutations in this gene have been associated with various behavioral phenotypes, including autonomic nervous system dysfunction, attention deficit/hyperactivity disorder, and the personality trait of novelty seeking. This gene contains a polymorphic number (2-10 copies) of tandem 48 nt repeats; the sequence shown contains four repeats.
Specifications
| D4 Dopamine Receptor | |
| Polyclonal | |
| Unconjugated | |
| DRD4 | |
| AW125663; D(2C) dopamine receptor; D(4) dopamine receptor; D4DR; D4R; D4RA; Dopamine 4 receptor; Dopamine D4 receptor; dopamine receptor 4; dopamine receptor D4; DRD4; Drd-4; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 13491, 1815 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P21917, P51436 | |
| DRD4 | |
| KLH conjugated synthetic peptide between 365-391 amino acids from the Central region of human DRD4. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction