missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DRD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533482
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gibbon, Monkey, Marmoset, Mouse, Dog, Bat, Hamster, Panda, Horse, Pig (100%).
This gene encodes the D3 subtype of the five (D1-D5) dopamine receptors. The activity of the D3 subtype receptor is mediated by G proteins which inhibit adenylyl cyclase. This receptor is localized to the limbic areas of the brain, which are associated with cognitive, emotional, and endocrine functions. Genetic variation in this gene may be associated with susceptibility to hereditary essential tremor 1. Alternative splicing of this gene results in transcript variants encoding different isoforms, although some variants may be subject to nonsense-mediated decay (NMD).
Specifications
| DRD3 | |
| Polyclonal | |
| Unconjugated | |
| DRD3 | |
| D(3) dopamine receptor; D3 receptor; D3DR; D3nf; D3R; Dopamine 3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopamine receptor D3; dopaminergic receptor D3; Drd3; essential tremor 1; ETM1; FET1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100061160, 100513049, 100769070, 13490, 1814, 487984 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P30728, P35462 | |
| DRD3 | |
| Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human DRD3 / Dopamine Receptor D3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Canine, Monkey, Hamster, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction