missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRD3 Polyclonal Antibody
Description
DRD3 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | DRD3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.65 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | DRD3 |
| Gene Accession No. | P19020, P30728 |
| Gene Alias | D(3) dopamine receptor; D3 receptor; D3DR; D3nf; D3R; Dopamine 3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopamine receptor D3; dopaminergic receptor D3; Drd3; essential tremor 1; ETM1; FET1 |
| Gene Symbols | DRD3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?