missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22022-1-AP-150UL
This item is not returnable.
View return policy
Description
The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.Specifications
| DRD2 | |
| Polyclonal | |
| Unconjugated | |
| DRD2 | |
| D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, DRD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13489, 1813 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14416, P61168 | |
| DRD2 | |
| DRD2 Fusion Protein Ag16821 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction