missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17934-1-AP-20UL
This item is not returnable.
View return policy
Description
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.Specifications
| DRD1 | |
| Polyclonal | |
| Unconjugated | |
| DRD1 | |
| D(1A) dopamine receptor, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13488, 1812 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21728, Q61616 | |
| DRD1 | |
| DRD1 Fusion Protein Ag12366 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction