missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRD1 Monoclonal Antibody (SG2-D1a), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA146024
This item is not returnable.
View return policy
Description
This antibody does not react with human, canine, porcine, sheep, rabbit, or bovine.
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
Specifications
| DRD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P18901, Q61616 | |
| DRD1 | |
| Recombinant rat D1a dopamine receptor (123 residues with 23 residues near the C-terminus).[Swiss-Prot# P18901] | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
| SG2-D1a | |
| Unconjugated | |
| DRD1 | |
| C030036C15Rik; D(1A) dopamine receptor; D1 receptor; D1a; D1A dopamine receptor; D1a receptor; DADR; Dopamine 1 receptor; Dopamine D1 receptor; dopamine receptor D1; dopamine receptor D1A; Dopamine-1A receptor; Drd1; Drd-1; Drd1a; G protein-coupled receptor DRD1; Gpcr15 | |
| Mouse | |
| Protein A | |
| RUO | |
| 13488, 24316 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction