missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Draxin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112162
This item is not returnable.
View return policy
Description
This product can be used in direct ELISA.
Chemorepulsive axon guidance protein required for the development of spinal cord and forebrain commissures. Acts as a chemorepulsive guidance protein for commissural axons during development. Able to inhibit or repel neurite outgrowth from dorsal spinal cord. Inhibits the stabilization of cytosolic beta-catenin (CTNNB1) via its interaction with LRP6, thereby acting as an antagonist of Wnt signaling pathway.
Specifications
| Draxin | |
| Polyclonal | |
| PBS with no preservative | |
| Q8NBI3 | |
| DRAXIN | |
| Recombinant Human DRAXIN protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| DRAXIN | |
| 2610109H07Rik; AGPA3119; C1orf187; Dorsal inhibitory axon guidance protein; dorsal repulsive axon guidance protein; Draxin; FLJ34999; MGC117222; Neucrin; neural tissue-specific cysteine-rich protein; PSEC0258; UNQ3119; UNQ3119/PRO10268 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 374946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction