missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DRAQ5™

Product Code. 18359400 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
200 μL
500 μL
Unit Size:
200µL
500µL
50µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359400 500 μL 500µL
18075844 50 μL 50µL
18031936 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18359400 Supplier Novus Biologicals™ Supplier No. NBP281129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Content And Storage Store at 4°C in the dark. Do not freeze.
Quantity 500 μL
For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy, Fluorescence Imaging
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.