missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAP1 Antibody (PCRP-DRAP1-1A12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24152H
This item is not returnable.
View return policy
Description
DRAP1 Monoclonal antibody specifically detects DRAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| DRAP1 | |
| Monoclonal | |
| HRP | |
| dr1-associated corepressor, Dr1-associated protein 1, DR1-associated protein 1 (negative cofactor 2 alpha), NC2-alphaNegative co-factor 2-alpha, negative cofactor 2 alpha | |
| Recombinant human DRAP1 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| PCRP-DRAP1-1A12 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10589 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction