missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DRAM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553711
This item is not returnable.
View return policy
Description
Immunogen sequence: ISLRVEANLH GLTLYDTAPC PINNERTRLL SRDI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 82%.
Specifications
| DRAM2 | |
| Polyclonal | |
| Unconjugated | |
| DRAM2 | |
| 2010305N14Rik; 2610318G18Rik; AI647667; CORD21; damage regulated autophagy modulator 2; DNA damage regulated autophagy modulator 2; DNA damage-regulated autophagy modulator protein 2; DNA-damage regulated autophagy modulator 2; DRAM2; PRO180; PSEC0031; RP5-1180E21.1; TMEM77; Transmembrane protein 77; UNQ154/PRO180; VDNA-damage regulated autophagy modulator 2; WWFQ154 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128338 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UX65 | |
| DRAM2 | |
| Recombinant protein corresponding to Human DRAM2. Recombinant protein control fragment (Product #RP-91259). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction