missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DRAM2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Specifications
Specifications
| Antigen | DRAM2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.49 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DRAM2 |
| Gene Accession No. | Q6UX65 |
| Gene Alias | 2010305N14Rik; 2610318G18Rik; AI647667; CORD21; damage regulated autophagy modulator 2; DNA damage regulated autophagy modulator 2; DNA damage-regulated autophagy modulator protein 2; DNA-damage regulated autophagy modulator 2; DRAM2; PRO180; PSEC0031; RP5-1180E21.1; TMEM77; Transmembrane protein 77; UNQ154/PRO180; VDNA-damage regulated autophagy modulator 2; WWFQ154 |
| Gene Symbols | DRAM2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
