missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DRAM Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828522
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828522 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828522 Supplier Invitrogen™ Supplier No. PA520335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20335 can be used with blocking peptide PEP-0455.

Damage-regulated autophagy modulator (DRAM) is a p53 target gene encoding a lysosomal protein that induces autophagy, a process that degrades cytosolic proteins and organelles. It has been suggested that activation of DRAM by p53 is simultaneous to the activation by p53 of one or more proapoptotic genes such as PUMA, Bax, etc., and that the signaling pathways regulated by these genes together promote a full cell death response. By itself, DRAM cannot induce apoptosis, but the fact that it is inactivated in certain cancers highlights the importance of DRAM and suggests that autophagy may play a more important role in cancer than initially suspected. At least two different isoforms of DRAM are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRAM
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene DRAM1
Gene Accession No. Q8N682, Q9DC58
Gene Alias 1200002N14Rik; Damage-regulated autophagy modulator; DNA damage regulated autophagy modulator 1; DNA damage-regulated autophagy modulator protein 1; DNA-damage regulated autophagy modulator 1; DRAM; Dram1; FLJ11259; hypothetical protein FLJ11259; zgc:101811
Gene Symbols DRAM1
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human DRAM.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55332, 679937, 71712
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.