missing translation for 'onlineSavingsMsg'
Learn More

DRAK2 Antibody, Novus Biologicals™

Product Code. 18350651 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350651 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350651 Supplier Novus Biologicals Supplier No. NBP176885SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DRAK2 Polyclonal specifically detects DRAK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRAK2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AB011421
Gene Alias DAP kinase-related apoptosis-inducing protein kinase 2, DRAK2death-associated protein kinase-related 2, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 17b, serine/threonine kinase 17b (apoptosis-inducing), serine/threonine-protein kinase 17B
Gene Symbols STK17B
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the carboxy terminus of human DRAK2. The immunogen is located within the last 50 amino acids of DRAK2. Amino Acid Squence: SKRFRFDDSLPNPHE
Purification Method Ion exchange chromatography
Quantity 0.025 mg
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9262
Test Specificity It has no cross responses to DAP or ZIP kinases. The approximately 70 kDa band is probably non-related to DRAK2 although it is peptide blockable.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.