missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DRAK2 Monoclonal antibody specifically detects DRAK2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DRAK2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 2C3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH16040 |
| Gene Alias | DAP kinase-related apoptosis-inducing protein kinase 2, DRAK2death-associated protein kinase-related 2, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 17b, serine/threonine kinase 17b (apoptosis-inducing), serine/threonine-protein kinase 17B |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?