missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dragosantol (>80%), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | alpha-Bisabolol |
| CAS | 515-69-5 |
| Molecular Formula | C15 H26 O |
| Quantity | 5 g |
| Synonym | 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(4-methyl-3-penten-1-yl)-, (αR,1R)-rel-, 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(4-methyl-3-pentenyl)-, (R*,R*)-, 3-Cyclohexene-1-methanol, α,4-dimethyl-α-(4-methyl-3-pentenyl)-, (αR,1R)-rel- (9CI), 5-Hepten-2-ol, 6-methyl-2-(4-methyl-3-cyclohexen-1-yl)- (6CI,7CI,8CI), rel-(αR,1R)-α,4-Dimethyl-α-(4-methyl-3-penten-1-yl)-3-cyclohexene-1-methanol, (±)-α-Bisabolol, Bisabolol, Camilol, Dragosantol, Dragosantol 100, Hydagen B, dl-α-Bisabolol, α-Bisabolol |
| InChI Formula | InChI=1S/C15H26O/c1-12(2)6-5-11-15(4,16)14-9-7-13(3)8-10-14/h6-7,14,16H,5,8-11H2,1-4H3/t14-,15+/m1/s1 |
| SMILES | CC(=CCC[C@](C)(O)[C@H]1CCC(=CC1)C)C |
| IUPAC Name | (2S)-6-methyl-2-[(1S)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol |
| Molecular Weight (g/mol) | 222.37 |
| Formula Weight | 222.1984 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction