Learn More
Draeger DraegerSensor XS Electrochemical Sensor
Product Code. 16411255
Change view
Click to view available options
For Use With (Equipment):
Draeger Portable Gas Detectors X-am 3000 and X-am 7000
Product Code. For Use With (Equipment)
16411255 Draeger Portable Gas Detectors X-am 3000 and X-am 7000
1 options
Product Code. 16411255 Supplier Draeger Supplier No. 6811365
  • ONLINE EXCLUSIVE PRICE
    £625.80
    £485.07 / Pack of 1
    Save £140.73
    22% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For portable gas detectors. Used for detecting many different gases and varpours. Long service life and low operating costs.
  • Monitor toxic gases and oxygen deficiency
  • Compatible with X-am 3000 and X-am 7000
  • Measuring range in vol -% and ppm
  • Simple to use due to Smart Technology
Specifications
Description Electrochemical sensor
For Use With (Equipment) Draeger Portable Gas Detectors X-am 3000 and X-am 7000
Type Gas Detection Sensor
For Use With (Application) Accurate Measurement of Multiple Gases, Including H2O21, HF1, HCI or Hydrazine, for Example