missing translation for 'onlineSavingsMsg'
Learn More

Draeger Short Term Tubes

Product Code. 15852385
Change view
Click to view available options
Components Measured:
Ethanol 100/a
Mercaptan 0.1/a
Nitrous Fumes 50/b
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
15852385 Nitrous Fumes 50/b -
10364020 Mercaptan 0.1/a Pack of 10
15887676 Ethanol 100/a Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15852385 Supplier Draeger Supplier No. 8103941
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easy to use Draeger tubes are proven to be very cost effective and reliable method for th measurement of gases.

  • Quick and targeted measurements
  • Easy handling either manually using the Draeger-Tube pump accuro or automatically with the Draeger X-act 5000
  • Field of application includes air examinations at the workplace, technical gas analysis in areas of emission/immission concentrations, measurement of compressed air/gases
  • Measures a large number of different gases and vapours

Specifications

Components Measured Nitrous Fumes 50/b
Type Short-term Draeger Sampling Tubes
For Use With (Application) Spot Measurement of More That 500 Different Gases
compliance-icons
Product Identifier
  • X10 Gas detector tubes Nitrous Fumes 50/b
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.