missing translation for 'onlineSavingsMsg'
Learn More

Draeger Draeger Nitrous Oxide Diffusion Tubes

Product Code. 15353686
Click to view available options
Components Measured:
Nitrous Oxide
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15353686

Brand: Draeger 8101472

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Draeger diffusion tubes are direct reading systems used for the determination of time weighted average concentrations over the duration of one to eight hours.

  • Easy to handle - used with a plastic holder that is attached to clothing
  • Measures very low contentrations
  • Measurement result can be read directly on the scale printed on the tube following the length of discolouration

Specifications

Components Measured Nitrous Oxide
For Use With (Equipment) Draeger Diffusion Tube Holder
Type Gas Detection Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.