missing translation for 'onlineSavingsMsg'
Learn More

Dracorhodin Perchlorate, TRC

Product Code. 30378114
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378114 25 mg 25mg
30381435 100 mg -
30382881 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378114 Supplier Toronto Research Chemicals Supplier No. TRCD67891025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125536-25-6
Molecular Formula C17 H15 O3 . Cl O4
Molecular Weight (g/mol) 366.75
Synonym 1-Benzopyrylium, 7-hydroxy-5-methoxy-6-methyl-2-phenyl-, perchlorate (1:1), 1-Benzopyrylium, 7-hydroxy-5-methoxy-6-methyl-2-phenyl-, perchlorate (salt) (9CI), Dracorhodin perchlorate
IUPAC Name 5-methoxy-6-methyl-2-phenylchromenylium-7-ol;perchlorate
SMILES COc1c(C)c(O)cc2[o+]c(ccc12)c3ccccc3.[O-]Cl(=O)(=O)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H14O3.ClHO4/c1-11-14(18)10-16-13(17(11)19-2)8-9-15(20-16)12-6-4-3-5-7-12;2-1(3,4)5/h3-10H,1-2H3;(H,2,3,4,5)
Formula Weight 366.0506
Chemical Name or Material Dracorhodin Perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.