missing translation for 'onlineSavingsMsg'
Learn More

DR6/TNFRSF21 Antibody (001), DyLight 405, Novus Biologicals™

Product Code. 18404617 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404617 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404617 Supplier Novus Biologicals Supplier No. NBP289313V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DR6/TNFRSF21 Monoclonal antibody specifically detects DR6/TNFRSF21 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DR6/TNFRSF21
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate DyLight 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human DR6/TNFRSF21 (Met 1-Leu 350; NP_055267.1) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 27242
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.